Specifications:- LC Application Compatibility: Preparative. LC Union Type: Adaptor. LC Connector Material: Stainless Steel. Port Siz e 1: 1/4-28. Port Size 2: 5/16-24. Port Type 1: Male. Port Type 2: Female. Technique: LC. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Preparative. LC Union Type: Adaptor. LC Connector Material: Stainless Steel. Port Siz e 1: 1/4-28. Port Size 2: 5/16-24. Port Type 1: Male. Port Type 2: Female. Technique: LC. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Preparative. LC Union Type: Adaptor. LC Connector Material: Stainless Steel. Port Siz e 1: 1/4-28. Port Size 2: 5/16-24. Port Type 1: Male. Port Type 2: Female. Technique: LC. 1 * 1 Pce HEWL5023-3093 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou