Specifications:- LC Application Compatibility: LC Preparative. LC Module Type Compatibility: Fraction Collector. LC Supplies Brand: InfinityLab. LC System Compatibility: 1290 Infinity II Purification system. LC Use With Instrument: G7158B G7159B. 1 * 3 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: LC Preparative. LC Module Type Compatibility: Fraction Collector. LC Supplies Brand: InfinityLab. LC System Compatibility: 1290 Infinity II Purification system. LC Use With Instrument: G7158B G7159B. 1 * 3 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: LC Preparative. LC Module Type Compatibility: Fraction Collector. LC Supplies Brand: InfinityLab. LC System Compatibility: 1290 Infinity II Purification system. LC Use With Instrument: G7158B G7159B. 1 * 3 Pce HEWL5043-1412 1 * 3 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou