Specifications:- Bottom Shape: U. LC Application Compatibility: Analytical. LC Supplies Brand: InfinityLab. Material(s)ofConstructio n: Polypropylene. Maximum Fill Volume: 1.2 mL. Number of Wells: 96. Technique: LC LC/MS. Well Plate Material: Polypro 1 * 25 Pce
Fournisseur: Agilent
Specifications:- Bottom Shape: U. LC Application Compatibility: Analytical. LC Supplies Brand: InfinityLab. Material(s)ofConstructio n: Polypropylene. Maximum Fill Volume: 1.2 mL. Number of Wells: 96. Technique: LC LC/MS. Well Plate Material: Polypro 1 * 25 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Bottom Shape: U. LC Application Compatibility: Analytical. LC Supplies Brand: InfinityLab. Material(s)ofConstructio n: Polypropylene. Maximum Fill Volume: 1.2 mL. Number of Wells: 96. Technique: LC LC/MS. Well Plate Material: Polypro 1 * 25 Pce HEWL5043-9308 1 * 25 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou