Specifications:- Hardware: SS. Inner Diameter (ID): 0.075 mm. LC Column Brand: AdvanceBio Peptide Mapping. LC Column Maximum Tempera ture: 60 C . LC Column Phase: C18. Length: 150 mm. Part Number Application: Peptide Mapping. Part Number Application 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Hardware: SS. Inner Diameter (ID): 0.075 mm. LC Column Brand: AdvanceBio Peptide Mapping. LC Column Maximum Tempera ture: 60 C . LC Column Phase: C18. Length: 150 mm. Part Number Application: Peptide Mapping. Part Number Application 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Hardware: SS. Inner Diameter (ID): 0.075 mm. LC Column Brand: AdvanceBio Peptide Mapping. LC Column Maximum Tempera ture: 60 C . LC Column Phase: C18. Length: 150 mm. Part Number Application: Peptide Mapping. Part Number Application 1 * 1 Pce HEWL5065-9925 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou