Specifications:- Filter Assembly Material: Stainless Steel. Inner Diameter (ID): 4.6 mm. LC Application Compatibility: Analytical. L C Supplies Brand: InfinityLab. Pore Size: 0.5 um. Pressure Limit: 1300 bar. Technique: LC. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Filter Assembly Material: Stainless Steel. Inner Diameter (ID): 4.6 mm. LC Application Compatibility: Analytical. L C Supplies Brand: InfinityLab. Pore Size: 0.5 um. Pressure Limit: 1300 bar. Technique: LC. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Filter Assembly Material: Stainless Steel. Inner Diameter (ID): 4.6 mm. LC Application Compatibility: Analytical. L C Supplies Brand: InfinityLab. Pore Size: 0.5 um. Pressure Limit: 1300 bar. Technique: LC. 1 * 1 Pce HEWL5067-1602 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou