Specifications:- Filter Assembly Material: Stainless Steel. Filter Material: Stainless Steel. Inner Diameter (ID): 4.6 mm. LC Applic ation Compatibility: Analytical. LC Module Type Compatibility: Autosampler Pump Column Compartment. LC Supplies Brand 1 * 5 Pce
Fournisseur: Agilent
Specifications:- Filter Assembly Material: Stainless Steel. Filter Material: Stainless Steel. Inner Diameter (ID): 4.6 mm. LC Applic ation Compatibility: Analytical. LC Module Type Compatibility: Autosampler Pump Column Compartment. LC Supplies Brand 1 * 5 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Filter Assembly Material: Stainless Steel. Filter Material: Stainless Steel. Inner Diameter (ID): 4.6 mm. LC Applic ation Compatibility: Analytical. LC Module Type Compatibility: Autosampler Pump Column Compartment. LC Supplies Brand 1 * 5 Pce HEWL5067-1612 1 * 5 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou