Specifications:- LC Application Compatibility: Bio-inert. LC Module Type Compatibility: Column Compartment Valve. LC Use With Instru ment: G1316C G7116A G7116B. Number of Ports: 13. Pressure Limit: 200 bar. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Bio-inert. LC Module Type Compatibility: Column Compartment Valve. LC Use With Instru ment: G1316C G7116A G7116B. Number of Ports: 13. Pressure Limit: 200 bar. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Bio-inert. LC Module Type Compatibility: Column Compartment Valve. LC Use With Instru ment: G1316C G7116A G7116B. Number of Ports: 13. Pressure Limit: 200 bar. 1 * 1 Pce HEWL5067-4159 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou