Specifications:- LC Application Compatibility: Analytical. LC Module Type Compatibility: Column Compartment Valve. LC Supplies Brand : InfinityLab. LC Use With Instrument: G7116B. LC Valve Material: Stainless Steel. Number of Ports: 18. Pressure Limi 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Analytical. LC Module Type Compatibility: Column Compartment Valve. LC Supplies Brand : InfinityLab. LC Use With Instrument: G7116B. LC Valve Material: Stainless Steel. Number of Ports: 18. Pressure Limi 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Analytical. LC Module Type Compatibility: Column Compartment Valve. LC Supplies Brand : InfinityLab. LC Use With Instrument: G7116B. LC Valve Material: Stainless Steel. Number of Ports: 18. Pressure Limi 1 * 1 Pce HEWL5067-4233 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou