Specifications:- Inner Diameter (ID): 0.17 mm. LC Application Compatibility: Analytical. LC Module Type Compatibility: Column Compar tment Valve. LC System Compatibility: 1260 Infinity II LC System 1260 Infinity II Prime LC System 1290 Infinity II LC 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Inner Diameter (ID): 0.17 mm. LC Application Compatibility: Analytical. LC Module Type Compatibility: Column Compar tment Valve. LC System Compatibility: 1260 Infinity II LC System 1260 Infinity II Prime LC System 1290 Infinity II LC 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Inner Diameter (ID): 0.17 mm. LC Application Compatibility: Analytical. LC Module Type Compatibility: Column Compar tment Valve. LC System Compatibility: 1260 Infinity II LC System 1260 Infinity II Prime LC System 1290 Infinity II LC 1 * 1 Pce HEWL5067-4300 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou