Specifications:- Capillary Material: Stainless Steel. Capillary Subtype: Quick Connect Capillary. Fitting Type: Quick Connect. Fitti ng Type Left End: Quick Connect Fitting. Fitting Type Right End: Special. Inner Diameter (ID): 0.075 mm. LC Supplies 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Material: Stainless Steel. Capillary Subtype: Quick Connect Capillary. Fitting Type: Quick Connect. Fitti ng Type Left End: Quick Connect Fitting. Fitting Type Right End: Special. Inner Diameter (ID): 0.075 mm. LC Supplies 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Material: Stainless Steel. Capillary Subtype: Quick Connect Capillary. Fitting Type: Quick Connect. Fitti ng Type Left End: Quick Connect Fitting. Fitting Type Right End: Special. Inner Diameter (ID): 0.075 mm. LC Supplies 1 * 1 Pce HEWL5067-6602 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou