Specifications:- Inner Diameter (ID): 0.17 mm. LC Application Compatibility: Bio-inert. LC Supplies Brand: InfinityLab. LC System Co mpatibility: 1260 Infinity II Bio-Inert LC System. Technique: LC. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Inner Diameter (ID): 0.17 mm. LC Application Compatibility: Bio-inert. LC Supplies Brand: InfinityLab. LC System Co mpatibility: 1260 Infinity II Bio-Inert LC System. Technique: LC. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Inner Diameter (ID): 0.17 mm. LC Application Compatibility: Bio-inert. LC Supplies Brand: InfinityLab. LC System Co mpatibility: 1260 Infinity II Bio-Inert LC System. Technique: LC. 1 * 1 Pce HEWL5067-6621 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou