Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Fraction Collector Autosampler. LC System Compatibility: 1290 Infinity II Purification system. LC Use With Instrument: G7158B. Technique: LC. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Fraction Collector Autosampler. LC System Compatibility: 1290 Infinity II Purification system. LC Use With Instrument: G7158B. Technique: LC. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Fraction Collector Autosampler. LC System Compatibility: 1290 Infinity II Purification system. LC Use With Instrument: G7158B. Technique: LC. 1 * 1 Pce HEWL5067-6668 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou