Specifications:- LC Application Compatibility: Bio-compatible. LC Module Type Compatibility: Valve. LC System Compatibility: 1290 in finity II Bio LC System. LC Use With Instrument: G7116B. LC Valve Material: MP35N. Number of Ports: 10. Pressure Limi 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Bio-compatible. LC Module Type Compatibility: Valve. LC System Compatibility: 1290 in finity II Bio LC System. LC Use With Instrument: G7116B. LC Valve Material: MP35N. Number of Ports: 10. Pressure Limi 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Bio-compatible. LC Module Type Compatibility: Valve. LC System Compatibility: 1290 in finity II Bio LC System. LC Use With Instrument: G7116B. LC Valve Material: MP35N. Number of Ports: 10. Pressure Limi 1 * 1 Pce HEWL5067-6682 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou