Specifications:- Capillary Material: PTFE. Capillary Subtype: Special. LC Application Compatibility: Preparative. LC System Compatib ility: 1290 Infinity II Purification system. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Material: PTFE. Capillary Subtype: Special. LC Application Compatibility: Preparative. LC System Compatib ility: 1290 Infinity II Purification system. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Material: PTFE. Capillary Subtype: Special. LC Application Compatibility: Preparative. LC System Compatib ility: 1290 Infinity II Purification system. 1 * 1 Pce HEWL5067-6802 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou