Specifications:- LC Application Compatibility: Preparative. LC Union Type: Adaptor. LC Use With Instrument: G1328C. LC Connector Mat erial: Stainless Steel. Port Size 1: 10-32 (Swagelock). Port Size 2: 5/16-24. Port Type 1: Female. Port Type 2: Male. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Preparative. LC Union Type: Adaptor. LC Use With Instrument: G1328C. LC Connector Mat erial: Stainless Steel. Port Size 1: 10-32 (Swagelock). Port Size 2: 5/16-24. Port Type 1: Female. Port Type 2: Male. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Preparative. LC Union Type: Adaptor. LC Use With Instrument: G1328C. LC Connector Mat erial: Stainless Steel. Port Size 1: 10-32 (Swagelock). Port Size 2: 5/16-24. Port Type 1: Female. Port Type 2: Male. 1 * 1 Pce HEWL5067-6845 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou