Specifications:- Inner Diameter (ID): 0.3 mm. LC Application Compatibility: Preparative. LC Component Type: Capillary. LC Supplies B rand: InfinityLab. LC System Compatibility: 1260 Infinity II Purification system 1290 Infinity II Purification system 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Inner Diameter (ID): 0.3 mm. LC Application Compatibility: Preparative. LC Component Type: Capillary. LC Supplies B rand: InfinityLab. LC System Compatibility: 1260 Infinity II Purification system 1290 Infinity II Purification system 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Inner Diameter (ID): 0.3 mm. LC Application Compatibility: Preparative. LC Component Type: Capillary. LC Supplies B rand: InfinityLab. LC System Compatibility: 1260 Infinity II Purification system 1290 Infinity II Purification system 1 * 1 Pce HEWL5067-7015 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou