Specifications:- LC Application Compatibility: Bio-inert. LC Module Type Compatibility: Valve. LC System Compatibility: 1260 Infinit y Bio-Inert LC System 1260 Infinity II Bio-Inert LC System. LC Use With Instrument: G1316C. LC Valve Material: Cerami 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Bio-inert. LC Module Type Compatibility: Valve. LC System Compatibility: 1260 Infinit y Bio-Inert LC System 1260 Infinity II Bio-Inert LC System. LC Use With Instrument: G1316C. LC Valve Material: Cerami 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Bio-inert. LC Module Type Compatibility: Valve. LC System Compatibility: 1260 Infinit y Bio-Inert LC System 1260 Infinity II Bio-Inert LC System. LC Use With Instrument: G1316C. LC Valve Material: Cerami 1 * 1 Pce HEWL5068-0095 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou