Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Autosampler. LC Use With Instrument: G7157 A. LC Valve Material: Stainless Steel. Number of Ports: 6. Pressure Limit: 600 bar. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Autosampler. LC Use With Instrument: G7157 A. LC Valve Material: Stainless Steel. Number of Ports: 6. Pressure Limit: 600 bar. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Autosampler. LC Use With Instrument: G7157 A. LC Valve Material: Stainless Steel. Number of Ports: 6. Pressure Limit: 600 bar. 1 * 1 Pce HEWL5068-0267 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou