Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Valve Column Compartment. LC Supplies Bran d: InfinityLab. LC System Compatibility: 1290 Infinity II Purification system. LC Valve Material: Stainless Steel. Nu 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Valve Column Compartment. LC Supplies Bran d: InfinityLab. LC System Compatibility: 1290 Infinity II Purification system. LC Valve Material: Stainless Steel. Nu 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Valve Column Compartment. LC Supplies Bran d: InfinityLab. LC System Compatibility: 1290 Infinity II Purification system. LC Valve Material: Stainless Steel. Nu 1 * 1 Pce HEWL5068-0331 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou