Specifications:- LC Application Compatibility: Bio-compatible. LC System Compatibility: 1290 infinity II Bio LC System. LC Use With Instrument: G7132A. LC Valve Material: PEEK. Number of Ports: 7. Pressure Limit: 1300 bar. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Bio-compatible. LC System Compatibility: 1290 infinity II Bio LC System. LC Use With Instrument: G7132A. LC Valve Material: PEEK. Number of Ports: 7. Pressure Limit: 1300 bar. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Bio-compatible. LC System Compatibility: 1290 infinity II Bio LC System. LC Use With Instrument: G7132A. LC Valve Material: PEEK. Number of Ports: 7. Pressure Limit: 1300 bar. 1 * 1 Pce HEWL5068-0377 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou