Cone supplies
Fournisseur: Agilent
5188-5359
HEWL5188-5359EA 156 EUR InStock
HEWL5188-5359
Cone supplies
Matériel de chromatographie Chromatographie, Pièces de rechange
A range of accessories and supplies for skimmer and sampling cones for use with our ICP-MS portfolio.
  • Viton O-ring for sampling cone that seals the sampling cone to the vacuum chamber
  • Screws for skimmer bases are available and used to install the sampling base to the vacuum chamber
  • They also provide round interface wrenches (in order to remove and install the skimmer cone) and sampling cone wrenches (used to remove and install the sampling cone)
High quality ICP-MS systems contain interfaces that comprise of a step-down vacuum stage, located between aMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité