Specifications:- Carbon Load: 18 %. Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Maximum Temperature: 60 C . LC Column Phase: C18. LC Platform: HPLC. Length: 250 mm. Particle Size: 5 um. Particle T 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Carbon Load: 18 %. Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Maximum Temperature: 60 C . LC Column Phase: C18. LC Platform: HPLC. Length: 250 mm. Particle Size: 5 um. Particle T 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Carbon Load: 18 %. Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Maximum Temperature: 60 C . LC Column Phase: C18. LC Platform: HPLC. Length: 250 mm. Particle Size: 5 um. Particle T 1 * 1 Pce HEWL518905-902 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou