5190-6897
HEWL5190-6897EA
136
EUR
InStock
HEWL5190-6897
Eau pour LC-MS
Eau
A portfolio of LC/MS grade solvents that provides best-in-class acetonitrile, methanol and water. With newer LC/MS instruments having lower detection limits and regulations requiring ppb and ppt analysis of compounds, it is important to have LC/MS grade solvents that does not increase baseline noise. These InfinityLab Ultrapure LC/MS solvents have been developed by Agilent R&D and manufactured to have one of the lowest amounts of overall organic and metal contaminants in the industry. Our use of clear borosilicate bottles and smaller volumes (1L bottles) also increases the lot-to-lot quality by reducing the number of contaminants leaching from the bottle.
- Uniquely Agilent – These solvents were developed and qualified with Agilent instrumentation to have the best-in-class lowest overall contaminants. Manufacturing specifications are best-in-class and approved by Agilent manufacturing and R&D.
- Time and Cost Savings – Reduce troubleshooting time and extends the life of columns with reduce contaminants.
- Quality – This solvent portfolio was manufactured under ISO 9001 and ISO 14001 certification. Our small-batch mentality and bottling procedures increases the lot-to-lot quality and reduces solvent contamination.
- Universal – Best used with high-end Agilent LCMS instrumentation (QQQ, QTOF, and TOF), but can be used with any LCMS instrument and vendor.