Specifications:- Hardware: SS. Inner Diameter (ID): 5.2 mm. LC Column Brand: Bio-Monolith Protein A. LC Column Maximum Temperature: 40 C . LC Column Phase: Protein A. LC Platform: Analytical - Stainless Steel, HPLC ( 600 bar). Length: 5 mm. Particle 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Hardware: SS. Inner Diameter (ID): 5.2 mm. LC Column Brand: Bio-Monolith Protein A. LC Column Maximum Temperature: 40 C . LC Column Phase: Protein A. LC Platform: Analytical - Stainless Steel, HPLC ( 600 bar). Length: 5 mm. Particle 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Hardware: SS. Inner Diameter (ID): 5.2 mm. LC Column Brand: Bio-Monolith Protein A. LC Column Maximum Temperature: 40 C . LC Column Phase: Protein A. LC Platform: Analytical - Stainless Steel, HPLC ( 600 bar). Length: 5 mm. Particle 1 * 1 Pce HEWL5190-6903 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou