Cap, 9 mm, blue, screw, bonded, PTFE/white silicone. Specifications:- Cap Color: Blue. Cap Material: Polypropylene. Certified: No. I ncludes Septa: No. Pre-Slit: No. Sample Containment Type: Caps & septa. Septa Layering: Bi-Layer. Septa Material: PTF 1 * 100 Pce
Fournisseur: Agilent
Cap, 9 mm, blue, screw, bonded, PTFE/white silicone. Specifications:- Cap Color: Blue. Cap Material: Polypropylene. Certified: No. I ncludes Septa: No. Pre-Slit: No. Sample Containment Type: Caps & septa. Septa Layering: Bi-Layer. Septa Material: PTF 1 * 100 Pce
COMMANDE
Développer tout / Réduire tout
Cap, 9 mm, blue, screw, bonded, PTFE/white silicone. Specifications:- Cap Color: Blue. Cap Material: Polypropylene. Certified: No. I ncludes Septa: No. Pre-Slit: No. Sample Containment Type: Caps & septa. Septa Layering: Bi-Layer. Septa Material: PTF 1 * 100 Pce HEWL5191-5618 1 * 100 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou