SPME Arrow, PDMS (polydimethylsiloxane), 1.10 mm, 100 um, red, 3/pk. Specifications:- Color Code: Red. Fiber/Arrow Phase: PDMS (Poly dimethylsiloxane). Outer Diameter (OD): 1.1 mm. Phase Thickness: 100 um. SPME Application: Volatiles. SPME Format: Ar 1 * 3 Pce
Fournisseur: Agilent
SPME Arrow, PDMS (polydimethylsiloxane), 1.10 mm, 100 um, red, 3/pk. Specifications:- Color Code: Red. Fiber/Arrow Phase: PDMS (Poly dimethylsiloxane). Outer Diameter (OD): 1.1 mm. Phase Thickness: 100 um. SPME Application: Volatiles. SPME Format: Ar 1 * 3 Pce
COMMANDE
Développer tout / Réduire tout
SPME Arrow, PDMS (polydimethylsiloxane), 1.10 mm, 100 um, red, 3/pk. Specifications:- Color Code: Red. Fiber/Arrow Phase: PDMS (Poly dimethylsiloxane). Outer Diameter (OD): 1.1 mm. Phase Thickness: 100 um. SPME Application: Volatiles. SPME Format: Ar 1 * 3 Pce HEWL5191-5862 1 * 3 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou