SPME Arrow, PDMS (polydimethylsiloxane), 1.50 mm, 250 um, black, 3/pk. Specifications:- Color Code: Black. Fiber/Arrow Phase: PDMS ( Polydimethylsiloxane). Outer Diameter (OD): 1.5 mm. Phase Thickness: 250 um. SPME Application: Volatiles. SPME Format 1 * 3 Pce
Fournisseur: Agilent
SPME Arrow, PDMS (polydimethylsiloxane), 1.50 mm, 250 um, black, 3/pk. Specifications:- Color Code: Black. Fiber/Arrow Phase: PDMS ( Polydimethylsiloxane). Outer Diameter (OD): 1.5 mm. Phase Thickness: 250 um. SPME Application: Volatiles. SPME Format 1 * 3 Pce
COMMANDE
Développer tout / Réduire tout
SPME Arrow, PDMS (polydimethylsiloxane), 1.50 mm, 250 um, black, 3/pk. Specifications:- Color Code: Black. Fiber/Arrow Phase: PDMS ( Polydimethylsiloxane). Outer Diameter (OD): 1.5 mm. Phase Thickness: 250 um. SPME Application: Volatiles. SPME Format 1 * 3 Pce HEWL5191-5867 1 * 3 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou