Specifications:- Cap Color: Clear. Cap Material: Polypropylene. Cap Size: 9 mm. Certified: Yes. Includes Septa: Yes. Manufacturer Co mpatibility: Agilent Perkin Elmer Waters Thermo Shimadzu Dionex. Pre-Slit: No. Sample Containment Type: Caps & septa. 1 * 100 Pce
Fournisseur: Agilent
Specifications:- Cap Color: Clear. Cap Material: Polypropylene. Cap Size: 9 mm. Certified: Yes. Includes Septa: Yes. Manufacturer Co mpatibility: Agilent Perkin Elmer Waters Thermo Shimadzu Dionex. Pre-Slit: No. Sample Containment Type: Caps & septa. 1 * 100 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Cap Color: Clear. Cap Material: Polypropylene. Cap Size: 9 mm. Certified: Yes. Includes Septa: Yes. Manufacturer Co mpatibility: Agilent Perkin Elmer Waters Thermo Shimadzu Dionex. Pre-Slit: No. Sample Containment Type: Caps & septa. 1 * 100 Pce HEWL5191-8151 1 * 100 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou