Specifications:- Filter Assembly Material: Titanium. Inner Diameter (ID): 4.6 mm. LC Application Compatibility: Bio-compatible. LC M odule Type Compatibility: Pump. LC System Compatibility: 1290 infinity II Bio LC System. LC Use With Instrument: G713 1 * 2 Pce
Fournisseur: Agilent
Specifications:- Filter Assembly Material: Titanium. Inner Diameter (ID): 4.6 mm. LC Application Compatibility: Bio-compatible. LC M odule Type Compatibility: Pump. LC System Compatibility: 1290 infinity II Bio LC System. LC Use With Instrument: G713 1 * 2 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Filter Assembly Material: Titanium. Inner Diameter (ID): 4.6 mm. LC Application Compatibility: Bio-compatible. LC M odule Type Compatibility: Pump. LC System Compatibility: 1290 infinity II Bio LC System. LC Use With Instrument: G713 1 * 2 Pce HEWL5320-0048 1 * 2 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou