Specifications:- Capillary Material: PEEK/SST. Capillary Subtype: Quick Connect Capillary. Fitting Type: Quick Connect. Fitting Type Left End: No fitting. Fitting Type Right End: No fitting. Inner Diameter (ID): 0.17 mm. LC Application Compatibility 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Material: PEEK/SST. Capillary Subtype: Quick Connect Capillary. Fitting Type: Quick Connect. Fitting Type Left End: No fitting. Fitting Type Right End: No fitting. Inner Diameter (ID): 0.17 mm. LC Application Compatibility 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Material: PEEK/SST. Capillary Subtype: Quick Connect Capillary. Fitting Type: Quick Connect. Fitting Type Left End: No fitting. Fitting Type Right End: No fitting. Inner Diameter (ID): 0.17 mm. LC Application Compatibility 1 * 1 Pce HEWL5500-1276 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou