Specifications:- Capillary Material: MP35N. Fitting Type Left End: SI. Fitting Type Right End: SI. Inner Diameter (ID): 0.25 mm. LC Application Compatibility: Bio-compatible. LC Component Type: Capillary. LC System Compatibility: 1290 infinity II Bio 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Material: MP35N. Fitting Type Left End: SI. Fitting Type Right End: SI. Inner Diameter (ID): 0.25 mm. LC Application Compatibility: Bio-compatible. LC Component Type: Capillary. LC System Compatibility: 1290 infinity II Bio 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Material: MP35N. Fitting Type Left End: SI. Fitting Type Right End: SI. Inner Diameter (ID): 0.25 mm. LC Application Compatibility: Bio-compatible. LC Component Type: Capillary. LC System Compatibility: 1290 infinity II Bio 1 * 1 Pce HEWL5500-1283 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou