Specifications:- Inner Diameter (ID): 0.8 mm. LC Application Compatibility: Preparative. LC Union Type: Adaptor. LC Connector Materi al: PEEK. Port Size 1: 1/4-28. Port Size 2: 10-32. Port Type 1: Female. Port Type 2: Male. Technique: LC. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Inner Diameter (ID): 0.8 mm. LC Application Compatibility: Preparative. LC Union Type: Adaptor. LC Connector Materi al: PEEK. Port Size 1: 1/4-28. Port Size 2: 10-32. Port Type 1: Female. Port Type 2: Male. Technique: LC. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Inner Diameter (ID): 0.8 mm. LC Application Compatibility: Preparative. LC Union Type: Adaptor. LC Connector Materi al: PEEK. Port Size 1: 1/4-28. Port Size 2: 10-32. Port Type 1: Female. Port Type 2: Male. Technique: LC. 1 * 1 Pce HEWL5500-1410 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou