Specifications:- Capillary Material: Stainless Steel. Capillary Subtype: Standard capillary. Fitting Type Left End: SX. Fitting Type Right End: SI. Inner Diameter (ID): 0.7 mm. LC Application Compatibility: Preparative. LC Component Type: Capillary. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Material: Stainless Steel. Capillary Subtype: Standard capillary. Fitting Type Left End: SX. Fitting Type Right End: SI. Inner Diameter (ID): 0.7 mm. LC Application Compatibility: Preparative. LC Component Type: Capillary. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Material: Stainless Steel. Capillary Subtype: Standard capillary. Fitting Type Left End: SX. Fitting Type Right End: SI. Inner Diameter (ID): 0.7 mm. LC Application Compatibility: Preparative. LC Component Type: Capillary. 1 * 1 Pce HEWL5500-1414 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou