Specifications:- Capillary Material: Stainless Steel. Capillary Subtype: Standard capillary. Fitting Type Left End: SX. Fitting Type Right End: No fitting. Inner Diameter (ID): 0.17 mm. LC Application Compatibility: Preparative Analytical. LC System 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Material: Stainless Steel. Capillary Subtype: Standard capillary. Fitting Type Left End: SX. Fitting Type Right End: No fitting. Inner Diameter (ID): 0.17 mm. LC Application Compatibility: Preparative Analytical. LC System 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Material: Stainless Steel. Capillary Subtype: Standard capillary. Fitting Type Left End: SX. Fitting Type Right End: No fitting. Inner Diameter (ID): 0.17 mm. LC Application Compatibility: Preparative Analytical. LC System 1 * 1 Pce HEWL5500-1423 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou