Specifications:- Capillary Material: PTFE. Inner Diameter (ID): 1.5 mm. LC Use With Instrument: G7158B. Outer Diameter (OD): 3.5 mm. Swage Status Fitting Left End: Pre-swaged. Swage Status Fitting Right End: Pre-swaged. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Material: PTFE. Inner Diameter (ID): 1.5 mm. LC Use With Instrument: G7158B. Outer Diameter (OD): 3.5 mm. Swage Status Fitting Left End: Pre-swaged. Swage Status Fitting Right End: Pre-swaged. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Material: PTFE. Inner Diameter (ID): 1.5 mm. LC Use With Instrument: G7158B. Outer Diameter (OD): 3.5 mm. Swage Status Fitting Left End: Pre-swaged. Swage Status Fitting Right End: Pre-swaged. 1 * 1 Pce HEWL5500-1429 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou