Specifications:- Bed Mass: 150 mg. Cartridge Material: Polypropylene. Cartridge Size: 6 mL. Compound Types: Strong Basic Compounds. Frit Material: Polyethylene. Frit Pore Size: 20 um. Primary Extraction Mechanism: Strong Cation Exchange. Sample Prep 1 * 30 Pce
Fournisseur: Agilent
Specifications:- Bed Mass: 150 mg. Cartridge Material: Polypropylene. Cartridge Size: 6 mL. Compound Types: Strong Basic Compounds. Frit Material: Polyethylene. Frit Pore Size: 20 um. Primary Extraction Mechanism: Strong Cation Exchange. Sample Prep 1 * 30 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Bed Mass: 150 mg. Cartridge Material: Polypropylene. Cartridge Size: 6 mL. Compound Types: Strong Basic Compounds. Frit Material: Polyethylene. Frit Pore Size: 20 um. Primary Extraction Mechanism: Strong Cation Exchange. Sample Prep 1 * 30 Pce HEWL5610-2035 1 * 30 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou