Specifications:- Bed Mass: 40 mg. Cartridge Material: Polypropylene. Cartridge Size: 1 mL. Compound Types: Lipids. Primary Extractio n Mechanism: Size Exclusion Hydrophobic Interactions. Sample Prep Brand: Bond Elut. Sample Prep Phase: Lipid Extracti 1 * 100 Pce
Fournisseur: Agilent
Specifications:- Bed Mass: 40 mg. Cartridge Material: Polypropylene. Cartridge Size: 1 mL. Compound Types: Lipids. Primary Extractio n Mechanism: Size Exclusion Hydrophobic Interactions. Sample Prep Brand: Bond Elut. Sample Prep Phase: Lipid Extracti 1 * 100 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Bed Mass: 40 mg. Cartridge Material: Polypropylene. Cartridge Size: 1 mL. Compound Types: Lipids. Primary Extractio n Mechanism: Size Exclusion Hydrophobic Interactions. Sample Prep Brand: Bond Elut. Sample Prep Phase: Lipid Extracti 1 * 100 Pce HEWL5610-2041 1 * 100 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou