Specifications:- Carbon Load: 10 %. Endcapped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 21.2 mm. LC Column Brand: ZORBAX. LC Column Maximum Temperature: 90 C up to pH 6 40 C for pH 6-8 . LC Column Phase: SB-C18. LC Platfo 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Carbon Load: 10 %. Endcapped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 21.2 mm. LC Column Brand: ZORBAX. LC Column Maximum Temperature: 90 C up to pH 6 40 C for pH 6-8 . LC Column Phase: SB-C18. LC Platfo 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Carbon Load: 10 %. Endcapped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 21.2 mm. LC Column Brand: ZORBAX. LC Column Maximum Temperature: 90 C up to pH 6 40 C for pH 6-8 . LC Column Phase: SB-C18. LC Platfo 1 * 1 Pce HEWL585150-902 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou