Specifications:- Carbon Load: 9 %. Endcapped: Double. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 21.2 mm. LC Col umn Brand: ZORBAX. LC Column Maximum Temperature: 60 C at pH 2-6 40 C at pH 6-9 . LC Column Phase: Eclipse Plus C18 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Carbon Load: 9 %. Endcapped: Double. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 21.2 mm. LC Col umn Brand: ZORBAX. LC Column Maximum Temperature: 60 C at pH 2-6 40 C at pH 6-9 . LC Column Phase: Eclipse Plus C18 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Carbon Load: 9 %. Endcapped: Double. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 21.2 mm. LC Col umn Brand: ZORBAX. LC Column Maximum Temperature: 60 C at pH 2-6 40 C at pH 6-9 . LC Column Phase: Eclipse Plus C18 1 * 1 Pce HEWL595150-902 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou