InfinityLab Poroshell 120 EC-C18, 2.1, 250 mm, 4 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 10 %. Endc apped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poros 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell 120 EC-C18, 2.1, 250 mm, 4 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 10 %. Endc apped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poros 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell 120 EC-C18, 2.1, 250 mm, 4 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 10 %. Endc apped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poros 1 * 1 Pce HEWL650750-902T 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou