Specifications:- Hardware: SS. Inner Diameter (ID): 3 mm. LC Column Brand: AdvanceBio Peptide Mapping. LC Column Maximum Temperature : 60 C . LC Column Phase: C18. Length: 100 mm. Part Number Application: Peptide Mapping. Part Number Application Doma 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Hardware: SS. Inner Diameter (ID): 3 mm. LC Column Brand: AdvanceBio Peptide Mapping. LC Column Maximum Temperature : 60 C . LC Column Phase: C18. Length: 100 mm. Part Number Application: Peptide Mapping. Part Number Application Doma 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Hardware: SS. Inner Diameter (ID): 3 mm. LC Column Brand: AdvanceBio Peptide Mapping. LC Column Maximum Temperature : 60 C . LC Column Phase: C18. Length: 100 mm. Part Number Application: Peptide Mapping. Part Number Application Doma 1 * 1 Pce HEWL655950-302 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou