Specifications:- Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 21.2 mm. LC Column Brand: InfinityLa b Poroshell 120. LC Column Maximum Temperature: 60 C . LC Column Phase: HPH-C18. LC Platform: Preparative HPLC. Lengt 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 21.2 mm. LC Column Brand: InfinityLa b Poroshell 120. LC Column Maximum Temperature: 60 C . LC Column Phase: HPH-C18. LC Platform: Preparative HPLC. Lengt 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 21.2 mm. LC Column Brand: InfinityLa b Poroshell 120. LC Column Maximum Temperature: 60 C . LC Column Phase: HPH-C18. LC Platform: Preparative HPLC. Lengt 1 * 1 Pce HEWL670050-702 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou