InfinityLab Poroshell 120 SB-C18, 3.0 x 250 mm, 4 um, with column ID. Specifications:- Carbon Load: 8 %. Endcapped: No. Guard Column : No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximu 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell 120 SB-C18, 3.0 x 250 mm, 4 um, with column ID. Specifications:- Carbon Load: 8 %. Endcapped: No. Guard Column : No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximu 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell 120 SB-C18, 3.0 x 250 mm, 4 um, with column ID. Specifications:- Carbon Load: 8 %. Endcapped: No. Guard Column : No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximu 1 * 1 Pce HEWL680970-302 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou