Argon gas filters for ICP-OES
Fournisseur: Agilent
6810010000
HEWL6810010000EA 279 EUR InStock
HEWL6810010000
Argon gas filters for ICP-OES
Filtres Filtres à gaz
These gas purifying cartridges for ICP-OES are a cost effective safety and preventative measure to assure the supply of the highest purity gas to the ICP-OES instrumentation.
  • Deliver clean gases for accurate analyzes
  • Both inlet and outlet connectors are equipped with stainless steel frits to prevent particulates from entering the gas stream
  • Easily replaced
Impure gases can cause installation delays, premature instrument failure, and flawed results. Purification is one of the most important steps you can take to optimize your ICP-OES system performance. Agilent brings the highest performance of gas purifiers to ICP-OES instrument users. We manufacture our own range of gas purifying cartridges specifically designed to remove impurity gases, moisture and hydrocarbons from the ICP-OES gas supplies. We recommend that each Agilent ICP-OES system uses a gas purification system with Agilent gas purifying cartridges. These systems ensure the highest quality gas and include a tag to remind you when replacement is required, so you can replace them before sensitivity decreases, results are impacted or the instrument suffers other failures.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité