InfinityLab Poroshell 120 SB-C18, 2.1 x 150 mm, 1.9 um, with column ID. Specifications:- Carbon Load: 8 %. Endcapped: No. Guard Colu mn: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Ma 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell 120 SB-C18, 2.1 x 150 mm, 1.9 um, with column ID. Specifications:- Carbon Load: 8 %. Endcapped: No. Guard Colu mn: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Ma 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell 120 SB-C18, 2.1 x 150 mm, 1.9 um, with column ID. Specifications:- Carbon Load: 8 %. Endcapped: No. Guard Colu mn: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Ma 1 * 1 Pce HEWL683675-902 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou