Specifications:- Endcapped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 80 C at pH 7 35 C at pH 2-12 . LC Column Phase: HILIC-Z. LC Platform: 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Endcapped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 80 C at pH 7 35 C at pH 2-12 . LC Column Phase: HILIC-Z. LC Platform: 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Endcapped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 80 C at pH 7 35 C at pH 2-12 . LC Column Phase: HILIC-Z. LC Platform: 1 * 1 Pce HEWL683675-924 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou