Specifications:- Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 80 C . LC Column Phase: SB-Aq. LC Platform: UHPLC Low Dispersion UHPLC. Length: 100 mm. Particle Size: 1. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 80 C . LC Column Phase: SB-Aq. LC Platform: UHPLC Low Dispersion UHPLC. Length: 100 mm. Particle Size: 1. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 80 C . LC Column Phase: SB-Aq. LC Platform: UHPLC Low Dispersion UHPLC. Length: 100 mm. Particle Size: 1. 1 * 1 Pce HEWL685675-914 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou