InfinityLab Poroshell 120 SB-Aq, 3.0 x 100 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Endcapped: No. Guar d Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshell 120. LC Colum 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell 120 SB-Aq, 3.0 x 100 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Endcapped: No. Guar d Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshell 120. LC Colum 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell 120 SB-Aq, 3.0 x 100 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Endcapped: No. Guar d Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poroshell 120. LC Colum 1 * 1 Pce HEWL685975-314T 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou