InfinityLab Poroshell 120 HILIC-Z, 2.1 x 50 mm, 2.7 um. Specifications:- Endcapped: No. Guard Column: No. Hardware: Conventional. In ner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 80 C at pH 7 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell 120 HILIC-Z, 2.1 x 50 mm, 2.7 um. Specifications:- Endcapped: No. Guard Column: No. Hardware: Conventional. In ner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 80 C at pH 7 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell 120 HILIC-Z, 2.1 x 50 mm, 2.7 um. Specifications:- Endcapped: No. Guard Column: No. Hardware: Conventional. In ner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 80 C at pH 7 1 * 1 Pce HEWL689775-924 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou