InfinityLab Poroshell 120 SB-Aq, 4.6 x 50 mm, 2.7 um, analytical LC column, with column ID. Specifications:- Endcapped: No. Guard Co lumn: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column 1 * 1 Pce
Fournisseur: Agilent
InfinityLab Poroshell 120 SB-Aq, 4.6 x 50 mm, 2.7 um, analytical LC column, with column ID. Specifications:- Endcapped: No. Guard Co lumn: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
InfinityLab Poroshell 120 SB-Aq, 4.6 x 50 mm, 2.7 um, analytical LC column, with column ID. Specifications:- Endcapped: No. Guard Co lumn: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column 1 * 1 Pce HEWL689975-914T 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou